Learn More
Cayman Chemical ANTIBODY PRN-1371 25mg
SDP
Supplier:  Cayman Chemical 3378725 MG
Encompass_Preferred
A triterpenoid saponin with anti-inflammatory and gastroprotective activities increases apoptosis of and decreases production of IL-1B and IL-6 in human MH7A rheumatoid arthritis fibroblast-like synoviocytes from 4-16 UM reduces gastric acid secretion and lesion size in rat models of hydrochloric acid and ethanol- aspirin- water immersion stress- or pyloric ligation-induced ulcer formation at 50 and 100 mg/kg
Catalog No. 50-294-7361
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.