Learn More
Cayman Chemical ANTIBODY DMPAC-Chol 1mg
SDP
Supplier:  Cayman Chemical 265831 MG
Encompass_Preferred
A potent Nampt inhibitor (IC50 5 nM) has antiproliferative activity in U251 glioblastoma HT-1080 fibrosarcoma PC3 prostate MiaPaCa2 pancreatic and HCT116 colon cancer cell lines (IC50s 1.8 2.1 2.7 7.4 and 2.0 nM respectively) led to rapid tumor regression in a U251 mouse xenograft model at a dose of 25 mg/kg once per day
Catalog No. 50-293-2662
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.