Learn More
Cayman Chemical ANTIBODY SKF 86002 5mg
SDP
Supplier:  Cayman Chemical 302025 MG
Encompass_Preferred
A dipeptide pan-caspase inhibitor inhibits caspase-1 (IC50 20 nM) caspase-3 (IC50 30 nM) and caspase-6 -7 -8 and -9 (IC50s 5-18 nM) selective for caspases over calpain I cathepsin B cathepsin D renin thrombin and Factor Xa (IC50s 10 M) inhibits anti-Fas antibody-induced caspase-3 processing and PARP cleavage in Jurkat cells at 0 05 M increases survival in a mouse model of anti-Fas antibody-induced liver failure at 0 25 mg/kg reduces infarct size in rat models of acute myocardial infarction-reperfusion or transient focal brain ischemia-reperfusion injury intravenously as a 20 mg/kg bolus dose followed by a 5 mg/kg per hour infusion
Catalog No. 50-298-8513
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.