Learn More
Bellco Glass, Inc. Useful for indirect fluorescent antibody test including test for toxoplasmosis. Black ceramic background with 8 rings 6mm in diameter. Material: ceramic
SDP
Supplier:  Bellco Glass, Inc. 563801940
Encompass_Preferred
Useful for indirect fluorescent antibody test including test for toxoplasmosis. Black ceramic background with 8 rings 6mm in diameter. Material: ceramic
Catalog No. 50-194-5825
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.