Learn More
Cayman Chemical ANTIBODY TCS 2210 500ug
SDP
Supplier:  Cayman Chemical 16142500 UG
Encompass_Preferred
A staurosporine analog that potently inhibits JAK2 kinase (IC50 1 nM) and downstream targets STAT5 (IC50 10-30 nM) and STAT3 in a human erythroleukemic cell line expressing the JAK2V617F mutation potently inhibits the epigenetic kinase PRK1 (PKN1) in vitro (IC50 8.6 nM)
Catalog No. 50-297-0430
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.