Learn More
Cayman Chemical RAD51-IN-17 25mg
SDP
Supplier:  Cayman Chemical 4225325 MG
Encompass_Preferred
An ionizable cationic lipid (pKa 6.38) has been used in the generation of LNPs for the delivery of siRNA intravenous administration of LNPs containing ATX-100 and encapsulating Factor VII siRNA decrease Factor VII blood levels in mice
Catalog No. 50-294-3682
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.