Learn More
Cayman Chemical 2AmIno2 ptolylacetIc AcI 5g
SDP
Supplier:  Cayman Chemical 422945 G
Encompass_Preferred
An mPGES-1 inhibitor inhibits PGE2 production in both SK-N-AS human and BV-2 mouse cells (IC50s 0.1 and 2 UM respectively) reduces LPS-induced increases in a variety of cytokines including CCL2 IL-6 and TNF-a in the mouse hippocampus but not kidney without affecting the mRNA expression of Cox2 at 10 mg/kg i.p.
Catalog No. 50-294-3551
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.