Learn More
Cayman Chemical ANTIBODY ZINC69391 10mg
SDP
Supplier:  Cayman Chemical 4186010 MG
Encompass_Preferred
A Mas receptor agonist binds to the Mas receptor in bovine aortic endothelial cell membranes and COS cells (IC50s 21 and 47.5 nM respectively) inhibits perivascular inflammation and increases plaque stability in an ApoE-/- mouse model of atherosclerosis at 0.58 Umol/kg per day inhibits the formation of angiotensin II-induced abdominal aortic aneurysm in ApoE-/- mice
Catalog No. 50-294-5828
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.