Learn More
Cayman Chemical ANTIBODYS-NIFE 5mg
SDP
Supplier:  Cayman Chemical 174315 MG
Encompass_Preferred
An inhibitor of IKCa1/KCa3.1 channels (IC50 9.4 nM) inhibits A23187-induced hyperpolarization of human mouse and rat erythrocytes (IC50s 14 15 and 9 nM respectively) selective over Nav and most Kv channels as well as 65 receptors and channels in a panel inhibits T-lymphocyte activation and cytokine release in vitro (IC50 ~50 nM) decreases colon inflammation and improves weight gain in a rat model of DNBS-induced IBD
Catalog No. 50-296-5461
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.