Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antide acetate, Thermo Scientific Chemicals

Catalog No. AAJ66692LB0
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66692LB0 5 mg
AAJ66692MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66692LB0 Supplier Thermo Scientific Chemicals Supplier No. J66692LB0
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112568-12-4
Molecular Formula C82H108ClN17O14
Molecular Weight (g/mol) 1591.32
MDL Number MFCD00133104,MFCD00133104
InChI Key QRYFGTULTGLGHU-NBERXCRTSA-N
Synonym antide, iturelix, antide acetate, nal-lys-gnrh, unii-94055uoq3y, nal-lys-gnrhant, d-alaninamide, n-acetyl-3-2-naphthalenyl-d-alanyl-4-chloro-d-phenylalanyl-3-3-pyridinyl-d-alanyl-l-seryl-n6-3-pyridinylcarbonyl-l-lysyl-n6-3-pyridinylcarbonyl-d-lysyl-l-leucyl-n6-1-methylethyl-l-lysyl-l-prolyl, n-acetyl-3-naphhalen-2-yl-d-alanyl-p-chloro-d-phenylalanyl-3-3-pyridyl-d-alanyl-l-seryl-n sup 6-nicotinyl-l-lysyl-n sup 6-nicotinyl-d-lysyl-l-leucyl-n sup 6-isopropyl-l-lysyl-l-prolyl-d-alaninamide, d-alaninamide, n-acetyl-3-2-naphthalenyl-d-alanyl-4-chloro-d-phenylalanyl-3-3-pyridinyl-d-alanyl-l-seryl-n sup 6-3-pyridinylcarbonyl-l-lysyl-n sup 6-3-pyridinylcarbonyl-d-lysyl-l-leucyl-n sup 6-1-methylethyl-l-lysyl-l-prolyl, iturelix usan:inn
PubChem CID 16130938
IUPAC Name (2S)-N-[(1R)-1-{[(1S)-1-{[(2S)-1-[(2S)-2-{[(1R)-1-carbamoylethyl]carbamoyl}pyrrolidin-1-yl]-1-oxo-6-[(propan-2-yl)amino]hexan-2-yl]carbamoyl}-3-methylbutyl]carbamoyl}-5-[(pyridin-3-yl)formamido]pentyl]-2-[(2S)-2-[(2R)-2-[(2R)-3-(4-chlorophenyl)-2-[(2R)-2-acetamido-3-(naphthalen-2-yl)propanamido]propanamido]-3-(pyridin-3-yl)propanamido]-3-hydroxypropanamido]-6-[(pyridin-3-yl)formamido]hexanamide
SMILES CC(C)C[C@H](NC(=O)[C@@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)[C@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)[C@H](CO)NC(=O)[C@@H](CC1=CC=CN=C1)NC(=O)[C@@H](CC1=CC=C(Cl)C=C1)NC(=O)[C@@H](CC1=CC=C2C=CC=CC2=C1)NC(C)=O)C(=O)N[C@@H](CCCCNC(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@H](C)C(N)=O

Specifications

Color White
Quantity 5 mg
Formula Weight 1591.3
Physical Form Solid
Chemical Name or Material Antide acetate
Hazard Category H360
Precautionary Statement P201-P202-P281-P308+P313-P501c
RTECSNumber AY2961700
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.