Learn More
STEMCELL Technologies Anti-Human CD3 Antibody, Clone UCHT1, APC, Size: 100 Tests
SDP
Supplier:  STEMCELL Technologies 60011AZ
Encompass_Preferred
Clone UCHT1 is a mouse (BALB/c) monoclonal antibody that reacts with the ~20 kDa CD3ε subunit of the human and chimpanzee T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ~95% of mature T cells and NKT cells, and variably on thymocytes. Verified for use with flow cytometry and cell separation assays, use this antibody in applications requiring the detection of CD3. This antibody clone has also been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human T Cell Isolation Kit (Catalog #19661), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851; partial blocking may be observed), EasySep™ HLA Whole Blood T Cell Enrichment Kit (Catalog #19951HLA) and EasySep™ HLA Whole Blood CD2 Positive Selection Kit (Catalog #18687HLA). Visit STEMCELL.com for more information and related products.
Catalog No. NC1667201
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.