Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix Scientific™ Antimicrobial Hand Sanitizer with Cap
SCI_ED

Catalog No. S27938 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Volume (Metric):
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S27938 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S27938 Supplier AquaPhoenix Scientific™ Supplier No. HS8200P

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antimicrobial hand sanitizer is intended for use as a no-rinse hand sanitizer and meets or exceeds the requirements of the USDA to be used as an E3 antimicrobial hand sanitizer.

  • Complies with the W.H.O. specifications and requirements for an effective hand sanitizer
  • Consistency similar to water
  • This high-quality hand sanitizer is a pH balanced formula that is a mild blend of skin emollients and alcohols and contains glycerine
  • Leaves hands feeling soft and moisturized
  • The product is mild on skin, yet tough enough to exceed FDA/USDA efficacy guidelines for E3 hand sanitizer and antiseptic hand treatments
  • Viscosity: 5 to 15 cSt
  • Poly bottle with flip cap

Specifications

Product Type Hand Sanitizer
Color Clear
Certifications/Compliance E3 Rated USDA
Composition 65% Ethanol
For Use With (Application) Hand Sanitizer
Packaging Plastic Spray Bottle
pH Range 5 to 8 pH
Volume (English) 16.91 oz.
Volume (Metric) 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.