Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(III) bromide, ultra dry, 99.999% (metals basis)

Catalog No. AA4373206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4373206 5 g
AA4373214 25 g
AA4373222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4373206 Supplier Thermo Scientific Chemicals Supplier No. 04373206
Add to Cart

CAS: 7789-61-9 | Br3Sb | 361.472 g/mol

Antimony(III) bromide is used in the synthesis of antimony compounds, in semiconductors and chemical analysis. It acts as a fire retardant in polyethylene polymers, as mordant and in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-61-9
Molecular Formula Br3Sb
Molecular Weight (g/mol) 361.472
MDL Number MFCD00016317
InChI Key RPJGYLSSECYURW-UHFFFAOYSA-K
Synonym antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
PubChem CID 24615
IUPAC Name tribromostibane
SMILES Br[Sb](Br)Br

Specifications

Boiling Point 280°C
Melting Point 96.6°C
Physical Form Lump
Quantity 5 g
UN Number UN3260
Merck Index 14,706
Solubility Information Soluble in alcohol,acetone,ammonia,benzene,chloroform,dilute hydrochloric acid,hydrobromic acid and carbon disulfide.
Formula Weight 361.48
Percent Purity 99.999%
Packaging Ampule
Refractive Index 1.74
Sensitivity Air Sensitive
Density 4.418 g/mL
Assay (metals basis)
Chemical Name or Material Antimony (III) bromide, Ultra dry
Show More Show Less
Hazard Category H302+H332-H335
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 232-179-8
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.