Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(III) iodide, ultra dry, 99.999% (metals basis)

Catalog No. AA4283109
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4283109 10 g
AA4283104 2 g
AA4283118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4283109 Supplier Thermo Scientific Chemicals Supplier No. 04283109
Only null left
Add to Cart

CAS: 7790-44-5 | I3Sb | 502.47 g/mol

Antimony(III) iodide is used as a dopant in the preparation of thermoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-44-5
Molecular Formula I3Sb
Molecular Weight (g/mol) 502.47
MDL Number MFCD00016322
InChI Key KWQLUUQBTAXYCB-UHFFFAOYSA-K
Synonym antimony iii iodide, stibine, triiodo, antimony triiodide, antimon iii iodid, antimony iodide sbi3, antimony iodide, antimony iii iodide, ultra dry, antimony iii triiodide, hsdb 437, triiodostibine #
PubChem CID 24630
IUPAC Name triiodostibane
SMILES [Sb+3].[I-].[I-].[I-]

Specifications

Boiling Point 420°C
Melting Point 168°C
Physical Form Powder
Quantity 10 g
UN Number UN3260
Merck Index 14,710
Solubility Information Soluble in benzene,alcohol,acetone,CS2,HCl,KI,SnCl4,C2H7N. Insoluble in CHCl3,and CCl4.Soluble in benzene,alcohol,acetone,carbondisulfide,hydrochloric acid,potassium iodide,tinterachloride,dimethylamine. Slightly soluble in ethanol and acetone. Insoluble in chloroform and carbonterachloride.
Formula Weight 502.46
Percent Purity 99.999%
Odor Acrid
Packaging Ampule
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Antimony (III) iodide, Ultra dry
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 232-205-8
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.