Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Potassium Tartrate Trihydrate (USP), Fisher Chemical™

Catalog No. A867250 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A867-250 250 g Glass Bottle
A867-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. A867-250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Chemical Identifiers

CAS 28300-74-5
Molecular Formula C8H4K2O12Sb2 · 3 H2O
MDL Number MFCD00167056
Synonym Tartrated Antimony

Specifications

Melting Point 100°C
Color White
pH 2.0
Physical Form Solid
Quantity 250 g
Assay Percent Range 99.0 to 103.0 %
Formula Weight 667.85
Percent Purity 99.0 to 103.0%
Grade USP
Identification Pass Test
Loss on Drying 2.7% max.
Packaging Glass Bottle
Chemical Name or Material Antimony Potassium Tartrate Trihydrate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.