Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™

Catalog No. sa450100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA450-100 100 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SA450-100
This item is not returnable. View return policy

Chemical Identifiers

CAS 10025-91-9 , 7647-01-0 , 7732-18-5
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name antimony(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Packaging Poly Bottle
Vapor Pressure 14mmHg at 20°C
Vapor Density 0.7g/mol
Density 1.0g/cm³
Physical Form Liquid
Concentration 990 to 1010 ppm
Color Colorless
pH 1
Quantity 100 mL
Grade Certified
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Chemical Name or Material Antimony, Reference Standard Solution, 1000ppm ±1%
Show More Show Less
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.