Learn More
Leinco Technologies Inc Anti-Mouse Thy1 (CD90) In Vivo PLATINUM; Clone T24/31; Isotype: Rat IgG2b κ
SDP
Supplier:  Leinco Technologies Inc C1789100 MG
Encompass_Preferred
Specificity: Anti-Mouse CD90 (CloneT24/31) reacts with mouse CD90. Antigen Distribution: CD90 is expressed by thymocytes, peripheral T cells, myoblasts, epidermal cells, and keratinocytes. Background: CD90 is a 25-35 kDa GPI-anchored protein belonging to the Ig superfamily. Applications: (WB, FA, WB, FC, IHC) CloneT24/31 is particularly useful for depletion of T lymphocytes. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-5857
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.