Learn More
Bioassay Systems QuantiChrom™ Antioxidant Assay Kit. For quantitative determination of total antioxidant capacity (TAC) and evaluation of drug effects on TAC metabolism.
SDP
Catalog No. 50489233 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of total antioxidant capacity (TAC) and evaluation of drug effects on TAC metabolism. Key Features: Sensitive and accurate. Use 20 µL sample. Linear detection range from 1.5 to 1000 µM Trolox equivalents. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 10 min. Can be readily automated as a high-throughput assay for thousands of samples per day. Method: OD570nm. Samples: Serum, plasma, urine, saliva, food and beverage etc. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 1.5 µM.
Catalog No. 50-489-233 Supplier Bioassay Systems Supplier No. DTAC100
For quantitative determination of total antioxidant capacity (TAC) and evaluation of drug effects on TAC metabolism. Key Features: Sensitive and accurate. Use 20 µL sample. Linear detection range from 1.5 to 1000 µM Trolox equivalents. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 10 min. Can be readily automated as a high-throughput assay for thousands of samples per day. Method: OD570nm. Samples: Serum, plasma, urine, saliva, food and beverage etc. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 1.5 µM.