Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Antiseptic Bio-Hand Cleaner

Catalog No. 19090850 Shop All Medique Products
Change view
Click to view available options
Quantity:
25 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-850 25 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-850 Supplier Medique Supplier No. 51173
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Instant hand sanitizer with fresh scent.

  • Contains Ethyl Alcohol 66.5%

Specifications

Active Ingredients Ethyl Alcohol 66.5%
For Use With (Application) Hand sanitizer
Description Antiseptic Bio-Hand Cleaner
Quantity 25 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.