Description
Offer exceptional durability
- Inherently antistatic, fluid impervious, polyethylene-based film
- Skid-resistant for added safety
- Available with seamless bottom or with elastic seamed bottom for higher skid resistance
Order Info
Packed in double polybag.
Specifications
Specifications
| Size | Regular |
| Material | Polyethylene |
| Closure Type | Elastic Opening |
| Sole Type | Seamed Bottom |
| Construction | Polyethylene |
| Resistance | Fluid |
| Product Type | Shoe Covers |