Learn More
Creative Biomart Crotalus atrox antivenin
Supplier:  Creative Biomart THP0160
Encompass_Preferred
The product is derived and purified immunoglobulin fragments obtained from other domestic animals such as sheep previously immunized with Crotalus atrox (Western Diamondback rattlesnake). The final purified antivenin product is produced by mixing other different monospecific snake antivenins and isolating the antivenin of interest through fractionation and chromatography techniques.
Catalog No. 50-208-4211
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.