Learn More
TARGETMOL CHEMICALS INC ANTROQUINONOL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T30087
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Antroquinonol is a ubiquinone derivative from Antrodia camphorata with anti-inflammatory and anticancer activities.Antroquinonol reduces oxidative stress and inhibits inflammation and sclerosis in mice with focal segmental glomerulosclerosis through enhancement of the Nrf2 signaling pathway.Antroquinonol has potential for use in the treatment of non-small cell lung cancer. Antroquinonol has potential for the treatment of non-small cell lung cancer and may be used in colon cancer research. purity: 99%
Catalog No. 50-258-0431
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.