Learn More
ANXA11 Mouse anti-Human, Clone: OTI1C6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500950

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56-kD antigen recognized by sera from patients with various autoimmune diseases. Transcript variants encoding the same isoform have been identified.Specifications
ANXA11 | |
Monoclonal | |
Unconjugated | |
ANXA11 | |
ALS23, ANX11, CAP-50, CAP50 | |
Mouse | |
Affinity Chromatography | |
RUO | |
311 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1C6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P50995 | |
ANXA11 | |
Full length human recombit protein of human ANXA11 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.