Learn More
Stoelting™ ANY-box Behavior Test System
SDP
Catalog No. 10000372 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
4-behavior test system
5-behavior test system
Size:
40 cm
100 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size
10-000-372 5-behavior test system 40 cm
10-000-378 4-behavior test system 100 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-372 Supplier Stoelting™ Supplier No. 65500
Only null left
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Behavior Test System is a completely automated system designed to make behavioral tracking convenient and affordable without compromising features.
  • At the heart of ANY-box is the ANY-maze software, coupled to Stoelting's ANY-maze interface (AMi-2) device.
  • This combination of core components allows you to automate as many as four ANY-boxes per digial interface for simultaneous testing.
Specifications
For Use With (Application) Neuroscience
Size 40 cm
Model 5-behavior test system
Includes 4 Boxes