Description
- At the heart of ANY-box is the ANY-maze software, coupled to Stoelting's ANY-maze interface (AMi-2) device.
- This combination of core components allows you to automate as many as four ANY-boxes per digial interface for simultaneous testing
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Size | 40 cm |
| Model | Sociability Box, Clear, Enclosure |