Learn More
Stoelting™ ANY-maze Cue Light
SDP
Catalog No. 10000599 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Blue
Green
Red
White
Yellow
Size:
10 mm
14 mm
22 mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size
10-000-599 Yellow 22 mm
10-000-595 Blue 22 mm
10-000-604 Green 10 mm
10-000-600 Green 14 mm
10-000-596 Green 22 mm
10-000-605 Red 10 mm
10-000-601 Red 14 mm
10-000-597 Red 22 mm
10-000-602 White 14 mm
10-000-598 White 22 mm
10-000-606 Yellow 10 mm
10-000-603 Yellow 14 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 10-000-599 Supplier Stoelting™ Supplier No. 60045Y
Add to Cart
Add to Cart
ANY-maze Cue lights are ideal for providing visible cues to animals during tests.
  • Lights are water resistant
  • Available in a variety of colors and sizes
  • Cue lights plug directly into the AMi-2 Digital interface
Specifications
For Use With (Application) Neuroscience
Size 22 mm
Model Yellow