Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Anza™ 10-Pack Starter Kit

Catalog No. IVGN3006
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IVGN3006 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IVGN3006 Supplier Invitrogen™ Supplier No. IVGN3006
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Anza™ 11 EcoRI, Anza™ 5 BamHI, Anza™ 1 NotI, Anza™ 16 HindIII, Anza™ 12 XbaI, Anza™ 3 BcuI, Anza™ 8 XhoI, Anza™ 10 DpnI, Anza™ 19 BglII, Anza™ 26 Eco32I, Anza™ T4 DNA Ligase Master Mix, Anz

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA substrates
  • Complete digestion in 15 min
  • Includes 10 common restriction enzymes
  • Complete with Anza T4 DNA Ligase Master Mix and Anza Alkaline Phosphatase

Certifications

CoA

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 20 μL each of Anza 11 EcoRI, Anza 5 BamHI, Anza 1 NotI, Anza 16 HindIII, Anza 12 XbaI
20 μL each of Anza 3 BcuI, Anza 8 XhoI, Anza 10 DpnI, Anza 19 BglII, Anza 26 Eco32I
100 μL Anza T4 DNA Ligase Master Mix
20 μL Anza Alkaline Phosphatase
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)

Store at -5 to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Methylation Sensitivity Methylation Sensitivities Varies by Enzyme
Enzyme HindIII, NotI, Alkaline Phosphatase, XhoI, BcuI, XbaI, DpnI, T4 DNA Ligase, Eco32I, BamHI, BglII, EcoRI
Compatible Buffer Anza™ 10X Red Buffer, Anza™ 10X Buffer
Type IIS RE No
Quantity 1 kit
Product Type Starter Kit
Research Category Traditional Cloning
Product Line Anza
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.