Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Anza™ 52 PvuII

Catalog No. IVGN0526
Encompass_Preferred
Change view
Click to view available options
Quantity:
4000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IVGN0526 4000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IVGN0526 Supplier Invitrogen™ Supplier No. IVGN0526
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA types
  • Complete digestion in 15 min
  • Overnight digestion without star activity

Certifications

CoA

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 800 μL Anza 52 PvuII
2 mL Anza Buffer (10X)
2 mL Anza Red Buffer (10X)

Store at -20°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme Pvu II
Compatible Buffer Anza™ 10X Red Buffer, Anza™ 10X Buffer
Sensitive to Heat Inactivation No
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 4000 units
Product Type Restriction Enzyme Cloning System
Research Category Traditional Cloning
Product Line Anza
Show More Show Less
Do Anza restriction enzymes contain BSA?

All Anza restriction enzymes contain BSA.

Are Anza enzymes stable during shipment if they freeze and thaw?

Enzymes may freeze during shipment on dry ice. This does not affect them as they have been tested and shown to be 100% active after three freeze-thaw cycles. For 24-48 hour delivery, enzymes may be shipped on ice pack as their performance is not affected by short exposure times at 4 degrees C.

Is it possible to perform a double digestion using an Anza restriction enzyme and a FastDigest restriction enzyme or Thermo Scientific restriction enzyme that is not available in the Anza collection?

For optimal results, we highly recommend exclusively using Anza restriction enzymes in double digestion as they all have 100% buffer compatibility. If a certain enzyme is not available in Anza format, but is available in FastDigest format or as a Thermo Scientific conventional restriction enzyme, it may be possible to perform a double digestion. For specific protocol recommendations, please contact our technical support with detailed information of the desired enzymes and DNA template.

Why does the Anza restriction enzyme come with a clear buffer and a red buffer?

When the next step following digestion is agarose gel electrophoresis, the Anza Red buffer can be used directly. The Anza Red buffer contains the necessary loading dye and density gradient for directly loading on to the agarose gel. The red dye does not interfere with digestion and helps reduce additional pipetting.

Use the clear Anza Buffer for applications that require analysis by fluorescence excitation, as the dyes in the Anza Red Buffer may interfere with some fluorescence measurements.

Why is there a number in the name of the Anza restriction enzymes?

Anza restriction enzymes contain a number preceding the name. This number provides the option to organize the enzymes in freezers numerically or alphabetically. More common enzymes tend to have a lower number.

What is star activity?

Star activity is the nonspecific cleavage of sequences that are similar to a recognition sequence, but not identical to the recognition sequence. This occurs when reaction conditions are suboptimal. Some restriction enzymes have more of a propensity for star activity than others. Factors that are included in suboptimal conditions that result in star activity include high glycerol concentration, high concentration of enzyme, prolonged incubation time, and use of the incorrect buffer.

Anza restriction enzymes in conjunction with the Anza buffer have been formulated to eliminate star activity when following the recommended protocol. Anza enzymes allow for the flexibility for complete digestion in as few as 15 minutes and longer reactions, up to 16 hours, can be safely digested without showing star activity.

What is the unit definition for Anza restriction enzymes?

- A unit is defined as the amount of enzyme required to completely digest 1 µg of substrate DNA in 50 µL of the reaction mixture in 1 hour at 37 degrees C.
- Anza restriction enzymes and buffer have been formulated such that 1 µL of Anza restriction enzyme cleaves 1 µg of substrate DNA in 15 minutes in Anza buffer.

What are isoschizomers?

Isoschizomers are restriction enzymes that have the same recognition sequence and the same specificity. The first restriction enzyme discovered is called the prototype and all subsequently identified enzymes that recognize that sequence are isoschizomers.

Can Anza restriction enzymes digest unpurified PCR products in PCR buffer?

-Digestions in several PCR reaction buffers were tested and the Anza restriction enzymes fully digested DNA. However, not all commercially available buffers could be tested.

To obtain the best results for unpurified PCR products, we recommend the following reaction conditions:
- Up to 10 µL of PCR reaction mixture
- 1 µL of 10X Anza Buffer
- 1 µL of Anza restriction enzyme
- Up to 20 µL of H2O
- 37 degrees C, 15 min

If digested PCR product will be used for cloning, it is necessary to purify the PCR fragment prior to digestion to remove the active polymerase. Active thermophilic DNA polymerase will still be present in the PCR mixture if it is not removed by purification and may alter the ends of the cleaved DNA thereby reducing ligation efficiency.

Are the Anza modifying enzymes compatible with the Anza Buffer?

- All Anza modifying enzymes are compatible with the Anza buffer.
- Anza Alkaline Phosphatase is supplied with the Anza buffer.
- The Anza T4 PNK Kit, Anza DNA End Repair Kit, and the Anza DNA Blunt End Kit include their own buffers which contain co-factors for full functionality of each. They all are compatible with the Anza Buffer.
- The Anza T4 DNA Ligase Master Mix comes with no additional buffer and is fully compatible with Anza buffer.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.