Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Anza™ T4 PNK Kit

Catalog No. IVGN2304
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IVGN2304 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IVGN2304 Supplier Invitrogen™ Supplier No. IVGN2304
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Anza™ T4 PNK Enzyme (Polynucleotide Kinase), Anza™ 10X T4 PNK Buffer,

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • Part of the Anza Restriction Enzyme Cloning System
  • Quick DNA phosphorylation in 15 minutes
  • Heat inactivated in 5 minutes

Certifications

CoA

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 50 reactions Anza T4 PNK Enzyme
0.5 mL Anza 10X T4 PNK Buffer

Store at -5 to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Enzyme Polynucleotide Kinase
Compatible Buffer Anza™ 1X Buffer, Anza™ 10X T4 PNK Buffer
Quantity 50 reactions
Product Type T4 PNK Kit
Product Line Anza

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.