Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AOC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104398 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104398 Supplier Invitrogen™ Supplier No. PA5104398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AOC3.

Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AOC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AOC3
Gene Accession No. O08590, O70423, Q16853
Gene Alias amine oxidase, copper containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Aoc3; Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; semicarbazide-sensitive amine oxidase; SSAO; VAP1; VAP-1; Vascular adhesion protein 1; VP97
Gene Symbols AOC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human AOC3(Accession Q16853), corresponding to amino acid residues T399-S449.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11754, 29473, 8639
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.