Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam AOPP Assay Kit, Plasma, Tissue, Cell Lysate, 200 Tests
Description
- Suitable for measuring advanced oxidation protein products (AOPP) in plasma, tissue homogenate, and cell lysate samples.
- Cited in 13 publications.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Format | Kit |
| Detection Method | Colorimetric |
| Detection Location | In-Solution Detection |
| No. of Reactions | 200 Tests |
| Target Molecule | Advanced Oxidation Protein Products |
| Sample Type | Plasma, Tissue Homogenate, Cell Lysate |
| Quantity | 200 Tests |
| For Use With (Application) | Assay AOPP in biofluids, cell/tissue extracts |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?