Learn More
ABclonal Technology AP0033-1000ul Phospho-ELK1-S38
SDP
Supplier:  ABclonal Technology AP00331000UL
Encompass_Preferred
This gene is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily Proteins of the TCF subfamily form a ternary complex by binding to the the serum response factor and the serum response element in the promoter of the c-fos proto-oncogene The protein encoded by this gene is a nuclear target for the ras-raf-MAPK signaling cascade This gene produces multiple isoforms by using alternative translational start codons and by alternative splicing Related pseudogenes have been identified on chromosomes 7 and 14
Catalog No. 50-001-11322
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.