Learn More
ABclonal Technology AP0165-500ul Phospho-NMDAR1-S8
SDP
Supplier:  ABclonal Technology AP0165500UL
Encompass_Preferred
The protein encoded by this gene is a critical subunit of N-methyl-D-aspartate receptors members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel These subunits play a key role in the plasticity of synapses which is believed to underlie memory and learning Cell-specific factors are thought to control expression of different isoforms possibly contributing to the functional diversity of the subunits Alternatively spliced transcript variants have been described
Catalog No. 50-001-11471
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.