Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP2 gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549862 Supplier Invitrogen™ Supplier No. PA549862
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total AP-2 gamma protein.

The protein encoded by this gene is a sequence-specific DNA-binding transcription factor involved in the activation of several developmental genes. The encoded protein can act as either a homodimer or heterodimer with other family members and is induced during retinoic acid-mediated differentiation. It plays a role in the development of the eyes, face, body wall, limbs, and neural tube.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2 gamma
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TFAP2C
Gene Accession No. Q61312, Q92754
Gene Alias AA409384; activating enhancer binding protein 2 gamma; activating enhancer-binding protein 2 gamma; activator protein 2 gamma; AP-2.2; AP2gamma; AP2-GAMMA; ERF1; estrogen receptor factor 1; hAP-2g; stimulated by retinoic acid gene 2; Stra2; Tcfap2c; TFAP2C; TFAP2G; Transcription factor AP-2 gamma; transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma); transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma); transcription factor AP-2, gamma; transcription factor ERF-1
Gene Symbols TFAP2C
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human AP2C (aa 401-450).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21420, 362280, 7022
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.