Learn More
AP2M1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1C8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503018

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The heterotetrameric coat assembly protein complex, also known as the adaptor-related protein complex 2 (AP-2), belongs to the adaptor complexes medium subunits family. The mu 1 subunit of the AP-2 complex (AP2M1) is required for the activity of a vacuolar ATPase, which is responsible for proton pumping occurring in the acidification of endosomes and lysosomes. AP2M1 has also been shown to associate with the HIV-1 protein Nef, suggesting that Nef may use AP-2 complex to enhance the rate of endocytosis of both CD4 and class I MHC. AP2M1 may also play an important role in regulating the intracellular trafficking and function of cytotoxic T-lymphocyte associated (CTLA)-4 protein. At least two isoforms of AP2M1 are known to exist.Specifications
AP2M1 | |
Monoclonal | |
Unconjugated | |
AP2M1 | |
AP50, CLAPM1, mu2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
116563, 1173, 11773, 607139 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1C8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P84091, P84092, Q96CW1 | |
AP2M1 | |
Human recombit protein fragment corresponding to amino acids 97-383 of human AP2M1 produced in E.coli. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.