Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP3D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567344
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567344 Supplier Invitrogen™ Supplier No. PA567344
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRVKAIRKFAV SQMSALLDSA HLLASSTQRN GICEVLYAAA WICGEFSEHL QEPHHTLEAM LRPRVTTLPG HIQAVYVQNV VKLYASILQQ KEQAG Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.

AP3D1 is a subunit of the AP3 adaptor-like complex, which is not associated with clathrin. The AP3D1 subunit is implicated in intracellular biogenesis and trafficking of pigment granules and possibly platelet dense granules and neurotransmitter vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP3D1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ap3d1
Gene Accession No. O14617
Gene Alias AA407035; adapter-related protein complex 3 subunit delta-1; adaptin, delta; adaptor related protein complex 3 delta 1 subunit; adaptor related protein complex 3 subunit delta 1; adaptor related protein complex 3, delta 1 subunit; adaptor-related protein complex 3 subunit delta-1; adaptor-related protein complex 3, delta 1 subunit; adaptor-related protein complex 3, delta subunit; adaptor-related protein complex AP-3, delta subunit; ADTD; ap3; AP-3 complex delta subunit, partial CDS; AP-3 complex subunit delta; AP-3 complex subunit delta-1; Ap3d; Ap3d1; Bolvr; delta adaptin; delta-adaptin; delta-adaptin, partial CDS; hBLVR; mBLVR1; mh; mocha; PRO0039; subunit of putative vesicle coat adaptor complex AP-3
Gene Symbols Ap3d1
Host Species Rabbit
Immunogen Recombinant Human AP3D1. Recombinant protein control fragment (Product #RP-107994).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8943
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.