Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP5B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564344
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564344 Supplier Invitrogen™ Supplier No. PA564344
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEGDGRLQPQ APSWPAAEEG EGERSLTARE HSPEEARELR AAVIQLLDTS YLLTPVAQAQ LLWLLGWALR GLQGQPPALF K Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 68%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen AP5B1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AP5B1
Gene Accession No. Q2VPB7
Gene Alias adaptor related protein complex 5 beta 1 subunit; adaptor related protein complex 5, beta 1 subunit; adaptor-related protein complex 5 beta subunit; adaptor-related protein complex 5, beta 1 subunit; AP-5; AP-5 complex subunit beta; AP-5 complex subunit beta-1; AP5B1; Beta5; PP1030
Gene Symbols AP5B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AP5B1. Recombinant protein control fragment (Product #RP-103797).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.