Learn More
Selleck Chemical LLC Apabetalone (RVX-208) 20mg 1044870-39-4 RVX-000222
SDP
Supplier:  Selleck Chemical LLC S729520MG
Encompass_Preferred
Apabetalone (RVX-208, RVX-000222) is a potent BET bromodomain inhibitor with IC50 of 0.510 ?M for BD2 in a cell-free assay, about 170-fold selectivity over BD1. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4609
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.