Learn More
Tonbo Bioscience Antibodies APC Anti-Human CD40 (G28.5) ..... Cat. No. 20-0410-T025 ..... Size 25 tests
Supplier:  Tonbo Bioscience 200410T025
Encompass_Preferred
Antibodies APC Anti-Human CD40 (G28.5) ..... Cat. No. 20-0410-T025 ..... Size 25 testsThe G28.5 antibody reacts with human CD40, a 45-48 kDa glycoprotein that is a member of the TNFReceptor superfamily. CD40 is expressed on B cells, follicular dendritic cells, macrophages, endothelial cells, and fibroblasts. CD40 functions as a receptor for CD154 (CD40 ligand) and mediates a wide variety of immune and inflammatory responses including co-stimulation, B cell differentiation, isotype class switching, and germinal center formation.
Catalog No. 50-210-2734
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.