Learn More
ABclonal Technology APC anti-Human CD90/Thy1
SDP
Supplier:  ABclonal Technology A26399100T
Encompass_Preferred
This gene encodes a cell surface glycoprotein and member of the immunoglobulin superfamily of proteins The encoded protein is involved in cell adhesion and cell communication in numerous cell types but particularly in cells of the immune and nervous systems The encoded protein is widely used as a marker for hematopoietic stem cells This gene may function as a tumor suppressor in nasopharyngeal carcinoma Alternative splicing results in multiple transcript variants
Catalog No. 50-000-09358
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.