Learn More
Tonbo Bioscience Antibodies APC Anti-Human IL-4 (MP4-25D2) ..... Cat. No. 20-7048-T025 ..... Size 25 tests
Supplier:  Tonbo Bioscience 207048T025
Encompass_Preferred
Antibodies APC Anti-Human IL-4 (MP4-25D2) ..... Cat. No. 20-7048-T025 ..... Size 25 testsThe MP4-25D2 antibody is specific for human IL-4, a pleiotropic cytokine that Th2 cell differentiation, B cell proliferation and differentiation, and B cell class switching to IgE. MP4-25D2 is a neutralizing antibody and is reported to cross react with rhesus monkey IL-4.
Catalog No. 50-210-2786
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.