Learn More
Tonbo Bioscience Antibodies APC Anti-Mouse CD209b (SIGN-R1) (22D1) ..... Cat. No. 20-2093-U025 ..... Size 25 µg
Supplier:  Tonbo Bioscience 202093U025
Encompass_Preferred
Antibodies APC Anti-Mouse CD209b (SIGN-R1) (22D1) ..... Cat. No. 20-2093-U025 ..... Size 25 µgThe 22D1 monoclonal antibody reacts with mouse CD209b, also known as SIGN-R1. CD209b is a single-pass type II transmembrane C-type lectin. SIGN-R1 is expressed on marginal zone macrophages in the spleen and on lymph node medullary macrophages. Mouse CD209b is one of five homologues to human CD209 (DC-SIGN) and functions to mediate the uptake of encapsulated organisms.
Catalog No. 50-210-2761
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.