Learn More
Cytek Biosciences Inc Antibodies APC Anti-Mouse CD272 (BTLA) (6A6) ..... Cat. No. 20-5954-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 205954U025
Encompass_Preferred
Antibodies APC Anti-Mouse CD272 (BTLA) (6A6) ..... Cat. No. 20-5954-U025 ..... Size 25 µgThe 6A6 antibody reacts with mouse CD272, a type 1 transmembrane glycoprotein and member of the Ig superfamily. CD272, also known as BTLA (B and T Lymphocyte Attenuator), is expressed on peripheral lymphocytes as well as developing B and T cells, macrophages, and dendritic cells. CD272 is a receptor for CD270 (HVEM) and is structurally similar to CD152 (CTLA-4) and CD279 (PD-1) and functions as a negative regulator of B and T cell responses.
Catalog No. 50-210-2781
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.