Learn More
Cytek Biosciences Inc Antibodies APC Anti-Mouse CD279 (PD-1) (RMP1-30) ..... Cat. No. 20-9981-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 209981U025
Encompass_Preferred
Antibodies APC Anti-Mouse CD279 (PD-1) (RMP1-30) ..... Cat. No. 20-9981-U025 ..... Size 25 µgThe RMP1-30 antibody is specific for mouse CD279, also known as programmed death-1 (PD-1), a 55 kDa glycoprotein member of the Ig superfamily of molecules. PD-1 exists in a monomeric form that is expressed by CD4- CD8- thymocytes, where it participates in the processes of clonal selection, elimination of autoreactive lymphocytes, and development of tolerance. PD-1 expression is also inducible upon activation of mature T cells, where it has been proposed to interact with the co-stimulatory receptor CD80 to limit T cell activation. Two ligands for PD-1, known as PD-L1 (B7-H1) and PD-L2 (B7-DC) are differentially expressed on T and B cells, monocytes, macrophages, NK cells or dendritic cells.
Catalog No. 50-210-2793
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.