Learn More
Cytek Biosciences Inc Antibodies APC Anti-Mouse Ly-6C (HK1.4) ….. Cat. No. 20-5932-U100 ….. Size 100 µg
Supplier:  Cytek Biosciences Inc 205932U100
Encompass_Preferred
Antibodies APC Anti-Mouse Ly-6C (HK1.4) ….. Cat. No. 20-5932-U100 ….. Size 100 µgThe HK1.4 monoclonal antibody reacts with mouse Ly-6C, a 15-17kD GPI-linked cell-surface protein. Ly-6C is expressed on monocyte/macrophage cells, endothelial cells, granulocytes and a subset of lymphocytes. Mice strains with the Ly-6.2 alloantigen have subsets of both Ly-6C positive CD4 and CD8 cells while Ly-6.1 expressing strains have only the CD8 Ly-6C positive subset.
Catalog No. 50-210-2779
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.