Learn More
Enterprise Technology Solutions APC BACK-UPS CS 350VA 120V
SDP
Supplier:  Enterprise Technology Solutions BK350
Encompass_Preferred
Protect your electronics with this 6-outlet, 120-volt battery backup system. 210-watt back-UPS features a surge energy rating of 1,020 joules. Serial connection lets you use your PC to access additional power protection and management features for your uninterruptible power supply (UPS) through a serial port. Disconnected battery notification warns you when a battery is not available to provide backup power. Cold-start capability provides temporary battery power when the utility power is out. Dataline surge protection protects connected equipment from power surges on the data lines. Hot-swappable batteries ensure clean, uninterrupted power to your protected equipment while the batteries are being replaced. Wiring fault indicator informs you of potentially dangerous wiring problems in the wall circuit.
Catalog No. NC2069981
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.