Learn More
Abcam APC/Cy5.5 Conjugation Kit, Lightning-Link, 100 μg
Description
- Labeling time: < 4 hr
- Hands-on time: 30 sec
- Antibody recovery: 100%
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Antibody |
| Shipping Condition | Ambient |
| Product Type | Protein Labeling Reagent |
| Content And Storage | Store at -20°C. |
| Format | Kit |
| Label or Dye | APC/Cy5.5 |
| Detection Method | Fluorescence |
| Quantity | 100 μg |
| For Use With (Application) | Flow cytometry, Immunocytochemistry |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.