Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam APC/Cy5.5 Conjugation Kit, Lightning-Link, 3x10 μg

Catalog No. 502585617
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-5617 3 x 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-5617 Supplier Abcam Supplier No. AB1028553X10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labeling in less than 4 hours with 100% antibody recovery and only 30 seconds of hands-on time.

  • Labeling time: less than 4 hr
  • Hands-on time: 30 sec
  • Antibody recovery: 100%

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Antibody
Label or Dye APC/Cy5.5
Shipping Condition Ambient
Product Type Protein Labeling Reagent
Content And Storage Store at -20°C.
Format Kit
Quantity 3 x 10 μg
For Use With (Application) Antibody conjugation, Flow cytometry, Immunocytochemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.