Learn More
Abcam APC/Cy7 Conjugation Kit, Lightning-Link, 1 mg
Description
- Labeling time: < 4 hr
- Hands-on time: 30 sec
- Antibody recovery: 100%
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Antibody |
| Format | Kit |
| Label or Dye | APC/Cy7 |
| Shipping Condition | Ambient |
| Product Type | Protein Labeling Reagent |
| Content And Storage | Store at -20°C. |
| Detection Method | Fluorescence |
| Quantity | 1 mg |
| For Use With (Application) | Flow Cytometry, Fluorescence Microscopy |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.