Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam APC/Cy7 Conjugation Kit, Lightning-Link, 3 x 10 μg

Catalog No. 502585613
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-5613 3 x 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-5613 Supplier Abcam Supplier No. AB1028593X10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides 100% antibody recovery with simple labeling in less than 4 hours and 30 seconds of hands-on time.

  • Labeling time: < 4 hr
  • Hands-on time: 30 sec
  • Antibody recovery: 100%

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Antibody
Shipping Condition Ambient
Label or Dye APC/Cy7
Format Kit
Product Type Protein Labeling Reagent
Detection Method Fluorescence
Quantity 3 x 10 μg
For Use With (Application) Flow Cytometry, Fluorescence Microscopy
Storage Requirements Store at -20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.